This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226991_m1
See other PTPRR GE Assays ›
Gene Symbol:
Ptprr
Gene Name:
protein tyrosine phosphatase, receptor type, R
Gene Aliases:
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213
Chromosome Location:
Chr.10: 116018200 - 116274932 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226991_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptprr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161837.1 NP_001155309.1 5 - 6 790 84
NM_001161838.1 NP_001155310.1 2 - 3 747 84
NM_001161839.1 NP_001155311.1 3 - 4 438 84
NM_001161840.1 NP_001155312.1 3 - 4 630 84
NM_011217.2 NP_035347.1 6 - 7 1396 84
GenBank mRNA AF041866.1 - 3 - 4 555 84
AF129509.1 - 2 - 3 672 84
BC119230.1 - 6 - 7 1329 8733979 84
BC120547.1 - 6 - 7 1329 40129470 84
D31898.1 - 6 - 7 1359 84
DV657487.1 - 3 - 4 84
Z30313.1 - 5 - 6 785 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001334.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptprr

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
negative regulation of epithelial cell migration
dephosphorylation
peptidyl-tyrosine dephosphorylation
ERBB2 signaling pathway
negative regulation of ERK1 and ERK2 cascade
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
hydrolase activity
phosphatase activity
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161838.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161838.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161839.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161839.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161840.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_001161840.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_011217.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 NM_011217.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000009963.mm.1 TC1000001334.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0