This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226419_m1
See other SLC30A6 GE Assays ›
Gene Symbol:
Slc30a6
Gene Name:
solute carrier family 30 (zinc transporter), member 6
Gene Aliases:
9530029F08Rik, ZnT6
Chromosome Location:
Chr.17: 74395608 - 74424229 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226419_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc30a6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252478.1 NP_001239407.1 3 - 4 233 145
NM_144798.6 NP_659047.2 3 - 4 233 130
GenBank mRNA AF233346.1 - 2 - 3 185 130
AK082907.1 - 3 - 4 210 130
AK160903.1 - 3 - 4 222 145
BC066162.1 - 3 - 4 200 30471027 130
BC068169.1 - 3 - 4 222 30542277 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001158.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc30a6

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
zinc II ion transport
Golgi to endosome transport
transmembrane transport
zinc II ion transmembrane transport
zinc ion transmembrane transporter activity
cation transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700010895.mm.1 NM_001252478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010895.mm.1 NM_001252478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010895.mm.1 NM_144798.6.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010895.mm.1 NM_144798.6 Refseq
Affymetrix Mouse Clariom S Array PSR1700010895.mm.1 TC1700001158.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0