This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226418_m1
See other SLC30A6 GE Assays ›
Gene Symbol:
Slc30a6
Gene Name:
solute carrier family 30 (zinc transporter), member 6
Gene Aliases:
9530029F08Rik, ZnT6
Chromosome Location:
Chr.17: 74395608 - 74424229 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226418_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc30a6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252478.1 NP_001239407.1 2 - 3 145 110
NM_144798.6 NP_659047.2 2 - 3 145 110
GenBank mRNA AF233346.1 - 1 - 2 97 110
AF395840.1 - 1 - 2 87 110
AK082907.1 - 2 - 3 122 110
AK150342.1 - 2 - 3 121 110
AK151120.1 - 2 - 3 121 110
AK160903.1 - 2 - 3 134 111
BC066162.1 - 2 - 3 112 30471027 110
BC068169.1 - 2 - 3 134 30542277 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001158.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc30a6

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
zinc II ion transport
Golgi to endosome transport
transmembrane transport
zinc II ion transmembrane transport
zinc ion transmembrane transporter activity
cation transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700010891.mm.1 NM_001252478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010891.mm.1 NM_001252478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010891.mm.1 NM_144798.6.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010891.mm.1 NM_144798.6 Refseq
Affymetrix Mouse Clariom S Array PSR1700010891.mm.1 TC1700001158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700010894.mm.1 NM_001252478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010894.mm.1 NM_001252478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010894.mm.1 NM_144798.6.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010894.mm.1 NM_144798.6 Refseq
Affymetrix Mouse Clariom S Array PSR1700010894.mm.1 TC1700001158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700010893.mm.1 NM_001252478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010893.mm.1 NM_001252478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010893.mm.1 NM_144798.6.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700010893.mm.1 NM_144798.6 Refseq
Affymetrix Mouse Clariom S Array PSR1700010893.mm.1 TC1700001158.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0