This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01225660_m1
See other SLC6A15 GE Assays ›
Gene Symbol:
Slc6a15
Gene Name:
solute carrier family 6 (neurotransmitter transporter), member 15
Gene Aliases:
AA536730, AI326450, AI326451, v7-3
Chromosome Location:
Chr.10: 103367458 - 103419379 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01225660_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc6a15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175328.3 NP_780537.1 1 - 2 345 62
GenBank mRNA AY149280.1 - 1 - 2 310 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY149280.1, NM_175328.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc6a15

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
neurotransmitter transport
amino acid transport
neutral amino acid transport
leucine transport
proline transport
proline transmembrane transport
sodium ion transmembrane transport
L-alpha-amino acid transmembrane transport
proline:sodium symporter activity
neurotransmitter:sodium symporter activity
symporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000009334.mm.1 NM_175328.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000009334.mm.1 NM_175328.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000009334.mm.1 TC1000001244.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 911717 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0