This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224863_m1
See other HEPH GE Assays ›
Gene Symbol:
Heph
Gene Name:
hephaestin
Gene Aliases:
C130006F04Rik, Cpl, mKIAA0698, sla
Chromosome Location:
Chr.X: 96455436 - 96574485 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224863_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Heph
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159627.1 NP_001153099.1 3 - 4 669 141
NM_001159628.1 NP_001153100.1 3 - 4 677 141
NM_010417.2 NP_034547.2 3 - 4 677 141
NM_181273.4 NP_851790.1 3 - 4 669 141
GenBank mRNA AF082567.1 - 3 - 4 789 141
AK032895.1 - 3 - 4 589 141
AK081330.1 - 3 - 4 645 141
AK086923.1 - 3 - 4 588 141
AK129194.1 - 3 - 4 625 141
BC048237.1 - 3 - 4 517 6474714 141
BC054442.1 - 3 - 4 517 6474713 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000919.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Heph

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
copper ion transport
iron ion transport
cellular iron ion homeostasis
erythrocyte differentiation
iron ion homeostasis
oxidation-reduction process
ferroxidase activity
copper ion binding
ferrous iron binding
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
metal ion binding
epoxyqueuosine reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_001159627.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_001159627.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_001159628.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_001159628.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_010417.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_010417.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_181273.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 NM_181273.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00005915.mm.1 TC0X00000919.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0