This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224329_m1
See other BORA GE Assays ›
Gene Symbol:
Bora
Gene Name:
bora, aurora kinase A activator
Gene Aliases:
6720463M24Rik, AI317232
Chromosome Location:
Chr.14: 99046247 - 99076450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224329_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bora
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175265.4 NP_780474.1 1 - 2 305 62
GenBank mRNA AK020138.1 - 1 - 2 463 62
AK034930.1 - 1 - 2 306 62
AK141389.1 - 1 - 2 256 62
AK167430.1 - 1 - 2 237 62
BC052183.1 - 1 - 2 206 6336038 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK034930.1, AK141389.1, AK167430.1, BC052183.1, NM_175265.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bora

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
regulation of mitotic nuclear division
regulation of protein localization
cell division
regulation of mitotic spindle organization
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009053.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009053.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009053.mm.1 TC1400001152.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400009052.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009052.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009052.mm.1 TC1400001152.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400009054.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009054.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009054.mm.1 TC1400001152.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400009055.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009055.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009055.mm.1 TC1400001152.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0