This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224313_m1
See other DIS3 GE Assays ›
Gene Symbol:
Dis3
Gene Name:
DIS3 mitotic control homolog (S. cerevisiae)
Gene Aliases:
2810028N01Rik
Chromosome Location:
Chr.14: 99076634 - 99099778 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224313_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dis3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028315.2 NP_082591.2 16 - 17 2229 97
GenBank mRNA AK032091.1 - 11 - 12 1319 97
AK038313.1 - 16 - 17 2098 97
BC027357.1 - 15 - 16 2114 4952415 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002600.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dis3

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
mRNA catabolic process
rRNA catabolic process
positive regulation of GTPase activity
CUT catabolic process
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
RNA binding
nuclease activity
endonuclease activity
exonuclease activity
5'-3' exoribonuclease activity
guanyl-nucleotide exchange factor activity
bidentate ribonuclease III activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019481.mm.1 NM_028315.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019481.mm.1 NM_028315.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019481.mm.1 TC1400002600.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0