This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222315_m1
See other ATP7B GE Assays ›
Gene Symbol:
Atp7b
Gene Name:
ATPase, Cu++ transporting, beta polypeptide
Gene Aliases:
Atp7a, WND, tx
Chromosome Location:
Chr.8: 21992783 - 22060074 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222315_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp7b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007511.2 NP_031537.2 14 - 15 3266 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001860.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp7b

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
copper ion transport
cellular copper ion homeostasis
cellular zinc ion homeostasis
lactation
copper ion import
intracellular copper ion transport
metal ion transport
copper ion transmembrane transport
response to copper ion
sequestering of calcium ion
copper ion export
nucleotide binding
copper-exporting ATPase activity
copper ion transmembrane transporter activity
copper ion binding
ATP binding
zinc ion binding
hydrolase activity
cation-transporting ATPase activity
copper-transporting ATPase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800014657.mm.1 NM_007511.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800014657.mm.1 NM_007511.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800014657.mm.1 TC0800001860.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0