This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220716_m1
See other PCID2 GE Assays ›
Gene Symbol:
Pcid2
Gene Name:
PCI domain containing 2
Gene Aliases:
A730042J05Rik
Chromosome Location:
Chr.8: 13077183 - 13105343 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220716_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcid2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178708.3 NP_848823.2 3 - 4 733 115
GenBank mRNA AK042945.1 - 3 - 4 223 115
AK045796.1 - 3 - 4 213 115
AK048722.1 - 3 - 4 224 115
AK082891.1 - 3 - 4 799 115
AK163697.1 - 3 - 4 220 115
BC006047.1 - 3 - 4 222 115
BC118533.1 - 3 - 4 733 40095328 115

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcid2

Gene Ontology Categories:

Function(s)

negative regulation of apoptotic process
regulation of mRNA stability
positive regulation of B cell differentiation
positive regulation of transcription, DNA-templated
spleen development
positive regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of cysteine-type endopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0