This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220657_m1
See other UNC5D GE Assays ›
Gene Symbol:
Unc5d
Gene Name:
unc-5 homolog D (C. elegans)
Gene Aliases:
D930029E11Rik, Unc5h4, mKIAA1777
Chromosome Location:
Chr.8: 28646717 - 29219682 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220657_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153135.3 NP_694775.1 15 - 16 2623 66
GenBank mRNA AJ487854.1 - 15 - 16 2624 66
AK220417.1 - 5 - 6 964 66
BC137615.1 - 15 - 16 2414 8860634 66
BC144784.1 - 13 - 14 2192 9053332 66
BC145695.1 - 12 - 13 2198 40130998 66
FJ824810.1 - 14 - 15 2502 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001964.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc5d

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
signal transduction
multicellular organism development
axon guidance
pyramidal neuron differentiation
regulation of neuron migration
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800015575.mm.1 NM_153135.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015575.mm.1 NM_153135.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015575.mm.1 TC0800001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800015576.mm.1 NM_153135.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015576.mm.1 NM_153135.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015576.mm.1 TC0800001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800015577.mm.1 NM_153135.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015577.mm.1 NM_153135.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015577.mm.1 TC0800001964.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0