This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219923_m1
See other SIM2 GE Assays ›
Gene Symbol:
Sim2
Gene Name:
single-minded homolog 2 (Drosophila)
Gene Aliases:
bHLHe15
Chromosome Location:
Chr.16: 94085198 - 94127253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219923_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sim2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011377.2 NP_035507.2 1 - 2 728 115
GenBank mRNA D63383.1 - 1 - 2 734 115
D64135.1 - 1 - 2 675 115
U40576.1 - 1 - 2 1060 115
U42554.1 - 1 - 2 400 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001053.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sim2

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
signal transduction
multicellular organism development
nervous system development
embryonic pattern specification
cell differentiation
lung development
negative regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008233.mm.1 NM_011377.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008233.mm.1 NM_011377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600008233.mm.1 TC1600001053.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600008234.mm.1 NM_011377.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008234.mm.1 NM_011377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600008234.mm.1 TC1600001053.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0