This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219209_m1
See other 4930557J02RIK GE Assays ›
Gene Symbol:
4930557J02Rik
Gene Name:
RIKEN cDNA 4930557J02 gene
Gene Aliases:
-
Chromosome Location:
Chr.5: 34118740 - 34135339 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219209_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930557J02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040703.1 2 - 3 812 96
GenBank mRNA AK016164.1 - 2 - 3 814 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DNA polymerase N Poln POL4P Anti-sense Chr. 5 - 34007179 - 34169526 on NCBI Build GRCm38
Downstream HAUS augmin-like complex,
subunit 3
Haus3 D4S43h,
D5H4S43,
D5H4S43E
19.5kb Anti-sense Chr. 5 - 34153896 - 34169424 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0