This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219093_m1
See other 1700113A16RIK GE Assays ›
Gene Symbol:
1700113A16Rik
Gene Name:
RIKEN cDNA 1700113A16 gene
Gene Aliases:
2210408E11Rik, AA408983
Chromosome Location:
Chr.3: 88171560 - 88177785 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219093_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700113A16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045997.1 1 - 2 195 87
GenBank mRNA AK007191.1 - 1 - 2 117 87
AK008852.1 - 1 - 2 193 87
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY708757.1 - 1 - 2 190 Chr. 3 - 88171625 - 88177782 [-] on Build GRCm38 87
GenBankmRNA BE381071.1 - 1 - 2 105 Chr. 3 - 88171741 - 88177695 [-] on Build GRCm38 87
GenBankmRNA BI853643.1 - 1 - 2 102 Chr. 3 - 88171556 - 88177692 [-] on Build GRCm38 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream myocyte enhancer factor 2D Mef2d C80750 -3.8kb Anti-sense Chr. 3 - 88141966 - 88172084 on NCBI Build GRCm38
Downstream expressed sequence AW047730 AW047730 23.9kb Anti-sense Chr. 3 - 88201586 - 88203484 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002301.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300017407.mm.1 NR_045997.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017407.mm.1 NR_045997.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017407.mm.1 TC0300002301.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017408.mm.1 NR_045997.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017408.mm.1 NR_045997.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017408.mm.1 TC0300002301.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017409.mm.1 NR_045997.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017409.mm.1 NR_045997.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017409.mm.1 TC0300002301.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0