This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218043_m1
See other MEF2D GE Assays ›
Gene Symbol:
Mef2d
Gene Name:
myocyte enhancer factor 2D
Gene Aliases:
C80750
Chromosome Location:
Chr.3: 88141966 - 88172084 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218043_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mef2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133665.3 NP_598426.1 7 - 8 1271 76
GenBank mRNA AK141270.1 - 7 - 8 1251 76
AK155778.1 - 7 - 8 1272 76
BC011070.1 - 7 - 8 1239 3497896 76
S68895.1 - 7 - 8 1090 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000716.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mef2d

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
endochondral ossification
chondrocyte differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
multicellular organism development
nervous system development
cell differentiation
skeletal muscle cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
enzyme binding
activating transcription factor binding
protein homodimerization activity
histone deacetylase binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300004916.mm.1 NM_133665.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300004916.mm.1 TC0300000716.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0