This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217445_m1
See other EPHA7 GE Assays ›
Gene Symbol:
Epha7
Gene Name:
Eph receptor A7
Gene Aliases:
Cek11, Ebk, Ehk3, Hek11, Mdk1
Chromosome Location:
Chr.4: 28813044 - 28967511 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217445_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epha7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122889.1 NP_001116361.1 3 - 4 1071 91
NM_001290434.1 NP_001277363.1 3 - 4 1085 91
NM_010141.4 NP_034271.3 3 - 4 1085 91
GenBank mRNA AK030480.1 - 3 - 4 1052 91
AK032973.1 - 3 - 4 890 91
AK033903.1 - 3 - 4 1086 91
AK049848.1 - 3 - 4 1052 91
BC026153.1 - 3 - 4 977 3991628 91
X79082.1 - 3 - 4 1064 91
X79083.1 - 3 - 4 1064 91
X79084.1 - 3 - 4 1064 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000234.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epha7

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
nervous system development
brain development
phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
retinal ganglion cell axon guidance
regulation of protein autophosphorylation
positive regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
branching morphogenesis of a nerve
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
regulation of ERK1 and ERK2 cascade
nephric duct morphogenesis
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ephrin receptor activity
GPI-linked ephrin receptor activity
protein binding
ATP binding
axon guidance receptor activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
chemorepellent activity
ephrin receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001445.mm.1 NM_001122889.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001445.mm.1 NM_001122889.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001445.mm.1 NM_010141.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400001445.mm.1 TC0400000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400001444.mm.1 NM_001122889.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001444.mm.1 NM_001122889.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001444.mm.1 NM_010141.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400001444.mm.1 TC0400000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400001446.mm.1 NM_001122889.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001446.mm.1 NM_001122889.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001446.mm.1 NM_010141.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400001446.mm.1 TC0400000234.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0