This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215757_s1
See other D330050I16RIK GE Assays ›
Gene Symbol:
D330050I16Rik
Gene Name:
RIKEN cDNA D330050I16 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 5388336 - 5390069 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215757_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D330050I16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033224.1 1 1682 68
GenBank mRNA AK089953.1 - 1 1223 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB775353.1 - 1 443 Chr. 19 - 5389559 - 5390071 [+] on Build GRCm38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream squamous cell carcinoma antigen recognized by T cells 1 Sart1 U5-110K Anti-sense Chr. 19 - 5377523 - 5389420 on NCBI Build GRCm38
Downstream RIKEN cDNA 4930481A15 gene 4930481A15Rik 16.7kb Sense Chr. 19 - 5406815 - 5422847 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK013846.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK084835.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK089953.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK160711.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 NR_033224.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK013846.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK084835.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK089953.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 AK160711.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364560 NR_033224.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128502 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK013846.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK084835.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK089953.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK160711.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 NR_033224.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK013846.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK084835.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK089953.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 AK160711.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364560 NR_033224.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128502 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116473 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128502 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10465011 - -
Affymetrix Mouse Gene 1.1 ST Array 10460580 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 692242 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 101170093 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1220200 - -
Illumina Sentrix mouse-6.v1 bead array 101170093 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101170093 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2547377 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0