This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215481_m1
See other AGPAT6 GE Assays ›
Gene Symbol:
Agpat6
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta)
Gene Aliases:
AU041707, AW545732, GPAT4, Tsarg7
Chromosome Location:
Chr.8: 23171265 - 23209171 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215481_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018743.4 NP_061213.2 3 - 4 1205 107
GenBank mRNA AF406611.1 - 2 - 3 716 107
AK045235.1 - 3 - 4 1099 107
AK049675.1 - 3 - 4 1099 107
AK077561.1 - 3 - 4 1089 107
AK083589.1 - 3 - 4 1099 107
AK161270.1 - 3 - 4 1068 107
AK167876.1 - 3 - 4 1114 107
AK169257.1 - 3 - 4 1099 107
AK182757.1 - 2 - 3 254 107
AY489184.1 - 2 - 3 274 107
BC031767.1 - 2 - 3 559 4457846 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001883.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agpat6

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glandular epithelial cell maturation
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
triglyceride metabolic process
phosphatidylcholine biosynthetic process
lactation
metabolic process
lipid biosynthetic process
phospholipid biosynthetic process
triglyceride biosynthetic process
mammary gland development
regulation of multicellular organism growth
diacylglycerol metabolic process
glycerol-3-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800014909.mm.1 NM_018743.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800014909.mm.1 NM_018743.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800014909.mm.1 TC0800001883.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0