This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212751_m1
See other TRIM46 GE Assays ›
Gene Symbol:
Trim46
Gene Name:
tripartite motif-containing 46
Gene Aliases:
TRIFIC
Chromosome Location:
Chr.3: 89234169 - 89246154 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212751_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trim46
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183037.2 NP_898858.1 1 - 2 182 76
GenBank mRNA AY251388.1 - 1 - 2 62 76
BC082299.1 - 1 - 2 182 76
CA320885.1 - 1 - 2 182 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002342.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trim46

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300017940.mm.1 NM_183037.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017940.mm.1 NM_183037.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300017940.mm.1 TC0300002342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017941.mm.1 NM_183037.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017941.mm.1 NM_183037.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300017941.mm.1 TC0300002342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017935.mm.1 NM_001039466.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017935.mm.1 NM_001039466.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017935.mm.1 NM_183037.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017935.mm.1 NM_183037.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300017935.mm.1 TC0300002342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017936.mm.1 NM_001039466.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017936.mm.1 NM_001039466.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017936.mm.1 NM_183037.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017936.mm.1 NM_183037.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300017936.mm.1 TC0300002342.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0