This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212466_g1
See other CD209D GE Assays ›
Gene Symbol:
Cd209d
Gene Name:
CD209d antigen
Gene Aliases:
SIGN-R3, SIGNR3, mSIGNR3
Chromosome Location:
Chr.8: 3871824 - 3878548 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212466_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd209d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130904.2 NP_570974.1 1 - 2 60 64
GenBank mRNA AF373411.1 - 1 - 2 60 64
AF440280.1 - 1 - 2 56 64
AK136586.1 - 1 - 2 116 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001623.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd209d

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
defense response to bacterium
positive regulation of cytokine secretion
mannose binding
carbohydrate binding
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800013350.mm.1 NM_130904.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800013350.mm.1 NM_130904.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800013350.mm.1 TC0800001623.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800013349.mm.1 NM_130904.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800013349.mm.1 NM_130904.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800013349.mm.1 TC0800001623.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0