This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212441_m1
See other CD209A GE Assays ›
Gene Symbol:
Cd209a
Gene Name:
CD209a antigen
Gene Aliases:
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, SIGN-R1, SIGNR5
Chromosome Location:
Chr.8: 3743395 - 3748984 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212441_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd209a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133238.5 NP_573501.1 6 - 7 640 79
GenBank mRNA AF373408.1 - 6 - 7 587 79
AF374470.1 - 5 - 6 506 79
AK054139.1 - 6 - 7 604 79
AK171639.1 - 6 - 7 584 79
AY049062.1 - 6 - 7 640 79
BC111095.1 - 5 - 6 688 40040036 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001619.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd209a

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
regulation of gene expression
regulation of blood coagulation
regulation of T cell proliferation
viral entry into host cell
virus receptor activity
mannose binding
carbohydrate binding
metal ion binding
calcium-dependent protein binding
carbohydrate derivative binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018061 AK171639.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018061 AY049062.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018061 AF373408.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018061 AK054139.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018061 NM_133238.5 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AK171639.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AY049062.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 BC111095.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AF373408.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AK054139.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AK171639.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AY049062.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 BC111095.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AF373408.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133884 AK054139.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638844 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584667 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370535 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116773 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AK171639.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AY049062.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 BC111095.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AF373408.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AK054139.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AK171639.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AY049062.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 BC111095.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AF373408.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133884 AK054139.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018061 AK171639.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018061 AY049062.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018061 AF373408.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018061 AK054139.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018061 NM_133238.5 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AK171639.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AY049062.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 BC111095.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AF373408.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AK054139.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AK171639.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AY049062.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 BC111095.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AF373408.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133884 AK054139.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615450 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615437 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615466 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615444 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615456 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615452 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615459 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615463 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615469 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615445 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615461 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615432 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615439 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615457 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615467 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615460 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615468 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116773 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06408241 - -
Whole Mouse Genome G4122A A_51_P133884 AK171639.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AY049062.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 BC111095.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AF373408.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AK054139.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AK171639.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AY049062.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 BC111095.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AF373408.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P133884 AK054139.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136320 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107777 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116773 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07073464 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07157531 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07157535 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07157537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615457 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615461 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615404 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615469 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615377 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615420 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615431 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615433 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615437 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615382 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615375 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615403 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07341403 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615457 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06744413 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615440 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615424 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615428 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615413 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07157534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615466 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07289894 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07427677 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615416 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615435 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615461 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06950199 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07151882 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615445 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07073464 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615411 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615430 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615401 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615463 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07144566 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615448 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615378 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615459 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615421 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615450 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615407 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615470 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615408 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615456 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615374 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07225554 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615453 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10576786 - -
Affymetrix Mouse Gene 1.0 ST Array 10576788 - -
Affymetrix Mouse Gene 1.0 ST Array 10576794 - -
Affymetrix Mouse Gene 1.0 ST Array 10576790 - -
Affymetrix Mouse Gene 1.0 ST Array 10576793 - -
Affymetrix Mouse Gene 1.0 ST Array 10576792 - -
Affymetrix Mouse Gene 1.0 ST Array 10576785 - -
Affymetrix Mouse Gene 1.0 ST Array 10576789 - -
Affymetrix Mouse Gene 1.0 ST Array 10576787 - -
Affymetrix Mouse Gene 1.0 ST Array 10576791 - -
Affymetrix HT MG-430 PM Array Plate 1426172_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10576784 - -
Mouse Genome 430 2.0 Array 1426172_PM_a_at - -
Mouse Genome 430 2.0 Array 1426172_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 170786_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 170786_at - -
Affymetrix Mouse Clariom S Array PSR0800013317.mm.1 NM_133238.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800013317.mm.1 NM_133238.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800013317.mm.1 TC0800001619.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800013318.mm.1 NM_133238.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800013318.mm.1 NM_133238.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800013318.mm.1 TC0800001619.mm.1 MTA TC
Mouse Expression Set 430 1426172_PM_a_at - -
Mouse Expression Set 430 1426172_a_at - -
Murine Genome U74v2 Set 134733_r_at - -
Murine Genome U74v2 Set 134733_PM_r_at - -
Mouse Genome 430A 2.0 Array 1426172_a_at - -
Mouse Genome 430A 2.0 Array 1426172_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1426172_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1426172_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 420071 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6590113 - -
Illumina mouse-6 v1.0 expression beadchip 6590113 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006590113 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617210 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2617210 - -
mouse-6 v1.1 (Illumina) scl35152.7.1_21-S - -
Illumina Sentrix mouse-6.v1 bead array 6590113 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl35152.7.1_21-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590113 - -
MouseRef-8 v2 Expression BeadChip ILMN_2617210 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617210 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0