This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210648_m1
See other MMP16 GE Assays ›
Gene Symbol:
Mmp16
Gene Name:
matrix metallopeptidase 16
Gene Aliases:
MT3-MMP, Mt3mmp
Chromosome Location:
Chr.4: 17853482 - 18118734 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210648_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmp16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019724.3 NP_062698.2 7 - 8 1463 108
GenBank mRNA AB021228.1 - 7 - 8 108
AF282844.1 - 7 - 8 1230 108
AK048543.1 - 7 - 8 1343 108
BC057926.1 - 7 - 8 1465 5292478 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000149.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mmp16

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

ossification
endochondral ossification
proteolysis
collagen catabolic process
chondrocyte proliferation
embryonic cranial skeleton morphogenesis
bone development
craniofacial suture morphogenesis
metalloendopeptidase activity
calcium ion binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400000971.mm.1 NM_019724.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400000971.mm.1 TC0400000149.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0