This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210364_m1
See other SMC4 GE Assays ›
Gene Symbol:
Smc4
Gene Name:
structural maintenance of chromosomes 4
Gene Aliases:
2500002A22Rik, C79747, SMC-4, Smc4l1
Chromosome Location:
Chr.3: 69004972 - 69034623 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210364_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133786.3 NP_598547.1 8 - 9 1237 145
GenBank mRNA AJ534940.2 - 7 - 8 1113 145
AK010848.1 - 6 - 7 688 145
AK048650.1 - 3 - 4 449 145
AK088350.1 - 8 - 9 1220 145
AK088846.1 - 8 - 9 1203 145
AK133817.1 - 8 - 9 1227 145
BC002078.1 - 3 - 4 304 145
BC061481.1 - 8 - 9 1212 6512832 145
BC062939.1 - 8 - 9 1237 6841276 145

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000574.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smc4

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
mitotic chromosome condensation
meiotic chromosome condensation
chromosome condensation
meiotic chromosome segregation
chromosome organization
cell division
kinetochore organization
nucleotide binding
protein binding
ATP binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300003864.mm.1 NM_133786.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003864.mm.1 NM_133786.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003864.mm.1 TC0300000574.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300003862.mm.1 NM_133786.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003862.mm.1 NM_133786.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003862.mm.1 TC0300000574.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300003869.mm.1 NM_133786.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003869.mm.1 NM_133786.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003869.mm.1 TC0300000574.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300003863.mm.1 NM_133786.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003863.mm.1 NM_133786.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003863.mm.1 TC0300000574.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300003867.mm.1 NM_133786.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003867.mm.1 NM_133786.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003867.mm.1 TC0300000574.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0