This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209969_m1
See other CCDC66 GE Assays ›
Gene Symbol:
Ccdc66
Gene Name:
coiled-coil domain containing 66
Gene Aliases:
E230015L20Rik
Chromosome Location:
Chr.14: 27481090 - 27508506 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209969_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccdc66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177111.3 NP_796085.3 15 - 16 2373 155
GenBank mRNA AK087583.1 - 7 - 8 1094 155
AK158666.1 - 13 - 14 1891 155
AK170006.1 - 13 - 14 1840 155
BC070471.1 - 15 - 16 2374 30626499 155
BC072583.1 - 7 - 8 1490 6403859 155

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001634.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccdc66

Gene Ontology Categories:

Function(s) Process(es)

retinal rod cell development
post-embryonic retina morphogenesis in camera-type eye
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400012322.mm.1 NM_177111.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400012322.mm.1 NM_177111.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400012322.mm.1 TC1400001634.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0