Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
NM_001109909.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
NM_001109909.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
NM_001109910.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
NM_001109910.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
NM_031405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
NM_031405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031197.mm.1 |
TC0500003439.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
NM_001109909.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
NM_001109909.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
NM_001109910.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
NM_001109910.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
NM_031405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
NM_031405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031195.mm.1 |
TC0500003439.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
NM_001109909.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
NM_001109909.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
NM_001109910.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
NM_001109910.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
NM_031405.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
NM_031405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031196.mm.1 |
TC0500003439.mm.1 |
MTA TC |