This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205687_g1
See other CPSF1 GE Assays ›
Gene Symbol:
Cpsf1
Gene Name:
cleavage and polyadenylation specific factor 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 76595808 - 76607613 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205687_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164173.1 NP_001157645.1 11 - 12 1362 100
NM_053193.2 NP_444423.1 11 - 12 1362 100
GenBank mRNA AF322193.1 - 11 - 12 1333 100
AK149914.1 - 4 - 5 362 100
AK155648.1 - 11 - 12 1303 100
AK155806.1 - 11 - 12 1284 100
BC011458.1 - 7 - 8 801 100
BC049941.1 - 11 - 12 1184 100
BC056388.1 - 11 - 12 1362 6854851 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001782.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf1

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA cleavage
mRNA processing
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500013744.mm.1 NM_001164173.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500013744.mm.1 NM_001164173.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500013744.mm.1 NM_053193.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500013744.mm.1 NM_053193.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500013744.mm.1 TC1500001782.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0