Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500015808.mm.1 |
NM_007984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015808.mm.1 |
NM_007984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015808.mm.1 |
TC0500001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015812.mm.1 |
NM_007984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015812.mm.1 |
NM_007984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015812.mm.1 |
TC0500001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015805.mm.1 |
NM_007984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015805.mm.1 |
NM_007984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015805.mm.1 |
TC0500001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015807.mm.1 |
NM_007984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015807.mm.1 |
NM_007984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015807.mm.1 |
TC0500001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015806.mm.1 |
NM_007984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015806.mm.1 |
NM_007984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015806.mm.1 |
TC0500001724.mm.1 |
MTA TC |