This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202101_m1
See other GCN1L1 GE Assays ›
Gene Symbol:
Gcn1l1
Gene Name:
GCN1 general control of amino-acid synthesis 1-like 1 (yeast)
Gene Aliases:
4932409G22, AL022764, G431004K08Rik, GCN1L, mKIAA0219
Chromosome Location:
Chr.5: 115565219 - 115622654 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202101_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcn1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172719.2 NP_766307.2 5 - 6 456 95
GenBank mRNA AK041717.1 - 5 - 6 445 95
AK090137.1 - 5 - 6 454 95
AK147257.1 - 5 - 6 449 95
AK154512.1 - 5 - 6 449 95
AK158040.1 - 5 - 6 447 95

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcn1l1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding
poly(A) RNA binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 620360 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0