This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201946_m1
See other SVOP GE Assays ›
Gene Symbol:
Svop
Gene Name:
SV2 related protein
Gene Aliases:
1110030H18Rik, AI415691, msvop
Chromosome Location:
Chr.5: 114026913 - 114091423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201946_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Svop
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026805.1 NP_081081.1 14 - 15 1543 76
GenBank mRNA AK029670.1 - 13 - 14 1252 76
AK047655.1 - 14 - 15 1536 76
AK047764.1 - 14 - 15 1544 76
AK053833.1 - 14 - 15 1534 76
AK082744.1 - 14 - 15 1545 76
BC059878.1 - 14 - 15 1496 6812082 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003055.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Svop

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transmembrane transport
transmembrane transport
ion transmembrane transporter activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500027175.mm.1 NM_026805.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027175.mm.1 NM_026805.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027175.mm.1 TC0500003055.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0