This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201440_m1
See other ERP29 GE Assays ›
Gene Symbol:
Erp29
Gene Name:
endoplasmic reticulum protein 29
Gene Aliases:
1200015M03Rik, 2810446M09Rik, AW209030, Erp28, Erp31, PDI-Db
Chromosome Location:
Chr.5: 121444753 - 121452474 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201440_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Erp29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026129.2 NP_080405.1 1 - 2 283 68
GenBank mRNA AB009293.1 - 1 - 2 160 68
AK004795.1 - 1 - 2 186 68
AK009881.1 - 1 - 2 189 68
AK154539.1 - 1 - 2 179 68
AK165702.1 - 183 68
AK168112.1 - 1 - 2 203 68
BC017125.1 - 1 - 2 283 4481563 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003188.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Erp29

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
positive regulation of protein phosphorylation
ER to Golgi vesicle-mediated transport
protein secretion
positive regulation of gene expression
negative regulation of gene expression
negative regulation of protein secretion
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
protein homodimerization activity
chaperone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500028382.mm.1 NM_026129.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028382.mm.1 NM_026129.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500028382.mm.1 TC0500003188.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500028379.mm.1 NM_026129.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028379.mm.1 NM_026129.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500028379.mm.1 TC0500003188.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500028381.mm.1 NM_026129.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028381.mm.1 NM_026129.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500028381.mm.1 TC0500003188.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0