This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201079_m1
See other STAMBPL1 GE Assays ›
Gene Symbol:
Stambpl1
Gene Name:
STAM binding protein like 1
Gene Aliases:
1700095N21Rik, 8230401J17Rik, ALMalpha, AMSH-FP
Chromosome Location:
Chr.19: 34192190 - 34240657 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201079_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stambpl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029682.4 NP_083958.3 3 - 4 658 74
GenBank mRNA AB010121.1 - 3 - 4 353 74
AB010122.1 - 3 - 4 317 74
AB066211.1 - 3 - 4 658 74
AB066213.1 - 3 - 4 341 74
AK085665.1 - 2 - 3 3049 74
AK138126.1 - 3 - 4 389 74
BC018343.1 - 3 - 4 384 4010423 74
BC023097.1 - 3 - 4 627 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000495.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stambpl1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
biological_process
molecular_function
peptidase activity
metallopeptidase activity
hydrolase activity
pyroglutamyl-peptidase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004419.mm.1 NM_029682.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004419.mm.1 NM_029682.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900004419.mm.1 TC1900000495.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0