This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200052_m1
See other MAGOHB GE Assays ›
Gene Symbol:
Magohb
Gene Name:
mago-nashi homolog B (Drosophila)
Gene Aliases:
2010012C16Rik, Mago, Magoh, Magoh-rs1
Chromosome Location:
Chr.6: 131284388 - 131293244 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200052_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Magohb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025564.2 NP_079840.2 2 - 3 230 136
GenBank mRNA AK006303.1 - 2 - 3 220 136
AK007769.1 - 2 - 3 217 136
AK008200.1 - 2 - 3 232 136
AK013310.1 - 2 - 3 215 136
BC027505.1 - 2 - 3 203 3372083 136
KF051005.1 - 2 - 3 151 136

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Magohb

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
transport
biological_process
RNA splicing
mRNA transport
RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0