This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01199518_m1
See other GNAI3 GE Assays ›
Gene Symbol:
Gnai3
Gene Name:
guanine nucleotide binding protein (G protein), alpha inhibiting 3
Gene Aliases:
AI158965, AW537698, Galphai3, Gnai-3
Chromosome Location:
Chr.3: 108107275 - 108149956 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01199518_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnai3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010306.3 NP_034436.1 7 - 8 1017 138
GenBank mRNA AK004566.1 - 7 - 8 1011 138
AK077490.1 - 7 - 8 979 138
AK133749.1 - 7 - 8 958 138
AK167438.1 - 7 - 8 971 138
AK168578.1 - 7 - 8 955 138
AK169067.1 - 7 - 8 962 138
AK169285.1 - 7 - 8 992 138
BC003462.1 - 7 - 8 943 138
BC020009.1 - 7 - 8 989 138
BC041107.1 - 7 - 8 928 2648164 138
U38502.1 - 1 - 2 131 138

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002731.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnai3

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

vesicle fusion
cell cycle
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
regulation of heart contraction
cell division
nucleotide binding
GTPase activity
signal transducer activity
protein binding
GTP binding
guanyl nucleotide binding
protein domain specific binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
GTPase activating protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300021412.mm.1 NM_010306.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021412.mm.1 TC0300002731.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0