This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01199475_m1
See other CUTC GE Assays ›
Gene Symbol:
Cutc
Gene Name:
cutC copper transporter homolog (E.coli)
Gene Aliases:
2310039I18Rik, AI326282, CGI-32
Chromosome Location:
Chr.19: 43753023 - 43768638 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01199475_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cutc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113562.1 NP_001107034.1 8 - 9 841 62
NM_025530.3 NP_079806.2 8 - 9 811 62
GenBank mRNA AK007591.1 - 8 - 9 766 62
AK009702.1 - 8 - 9 836 62
BC026775.1 - 8 - 9 751 3582902 62
BP754125.1 - 1 - 2 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000635.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cutc

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

protein tetramerization
copper ion homeostasis
copper ion binding
metal ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32128432 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32128433 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32128434 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 BP754125.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 NM_001113562.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 NM_025530.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 AK007591.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 AK009702.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 BP754125.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 NM_001113562.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 NM_025530.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 AK007591.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295582 AK009702.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511095 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710016 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509427 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815761 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298481 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531076 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397279 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918747 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633050 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118088 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157411 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175824 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 BP754125.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 NM_001113562.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 NM_025530.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 AK007591.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 AK009702.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 BP754125.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 NM_001113562.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 NM_025530.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 AK007591.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295582 AK009702.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295575 AK009702.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 BP754125.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 NM_001113562.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 NM_025530.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 AK007591.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 AK009702.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 BP754125.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 NM_001113562.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 NM_025530.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 AK007591.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295582 AK009702.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128446 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128445 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32128432 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157411 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03602059 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175824 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09179377 - -
Whole Mouse Genome G4122A A_51_P295582 BP754125.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 NM_001113562.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 NM_025530.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 AK007591.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 AK009702.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 BP754125.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 NM_001113562.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 NM_025530.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 AK007591.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295582 AK009702.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P295575 AK009702.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113039 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106077 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175824 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157411 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14477569 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14422222 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14477563 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32128433 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32128432 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32128434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128540 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128554 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128576 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128474 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128462 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128549 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128565 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32128519 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128435 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128432 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128493 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14441635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128448 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128443 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128438 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128489 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128459 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128494 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128436 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128497 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128445 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128439 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128504 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14432227 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128500 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128498 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14246505 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128454 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128449 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128453 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128452 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128501 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128502 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128490 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128461 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14448214 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128440 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14204655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14269163 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128447 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128446 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128458 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32128496 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10463304 - -
Affymetrix Mouse Gene 1.0 ST Array 10463305 - -
Affymetrix Mouse Gene 1.0 ST Array 10463307 - -
Affymetrix Mouse Gene 1.0 ST Array 10463301 - -
Affymetrix Mouse Gene 1.0 ST Array 10463306 - -
Affymetrix Mouse Gene 1.0 ST Array 10463299 - -
Affymetrix Mouse Gene 1.0 ST Array 10463302 - -
Affymetrix Mouse Gene 1.0 ST Array 10463298 - -
Affymetrix Mouse Gene 1.0 ST Array 10463300 - -
Affymetrix Mouse Gene 1.0 ST Array 10463297 - -
Affymetrix Mouse Gene 1.0 ST Array 10463303 - -
Affymetrix HT MG-430 PM Array Plate 1418473_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10463296 - -
Mouse Genome 430 2.0 Array 1418473_at - -
Mouse Genome 430 2.0 Array 1418473_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66388_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66388_PM_at - -
Affymetrix Mouse Clariom S Array PSR1900005642.mm.1 NM_001113562.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005642.mm.1 NM_001113562.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005642.mm.1 NM_025530.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005642.mm.1 NM_025530.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900005642.mm.1 TC1900000635.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900005641.mm.1 NM_001113562.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005641.mm.1 NM_001113562.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005641.mm.1 NM_025530.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005641.mm.1 NM_025530.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900005641.mm.1 TC1900000635.mm.1 MTA TC
Mouse Expression Set 430 1418473_at - -
Mouse Expression Set 430 1418473_PM_at - -
Mouse Genome 430A 2.0 Array 1418473_PM_at - -
Mouse Genome 430A 2.0 Array 1418473_at - -
Affymetrix HT Mouse Genome 430A Array 1418473_at - -
Affymetrix HT Mouse Genome 430A Array 1418473_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 859321 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 110452 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000110452 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2686087 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2686087 - -
mouse-6 v1.1 (Illumina) scl000477.1_15-S - -
Illumina Sentrix mouse-6.v1 bead array 110452 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl000477.1_15-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 110452 - -
MouseRef-8 v2 Expression BeadChip ILMN_2686087 - -
MouseRef-8 v2 Expression BeadChip ILMN_2834370 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2686087 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2834370 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000477.1_15-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0