This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01196532_m1
See other CAMKK2 GE Assays ›
Gene Symbol:
Camkk2
Gene Name:
calcium/calmodulin-dependent protein kinase kinase 2, beta
Gene Aliases:
6330570N16Rik, AW061083, mKIAA0787
Chromosome Location:
Chr.5: 122718580 - 122779476 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01196532_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camkk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199676.1 NP_001186605.1 11 - 12 1379 84
NM_145358.2 NP_663333.1 11 - 12 1379 84
GenBank mRNA AK031399.1 - 11 - 12 1304 84
AK032070.1 - 11 - 12 1352 84
AK044660.1 - 11 - 12 1368 84
AK122370.1 - 6 - 7 545 84
BC023103.1 - 10 - 11 1212 3993474 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camkk2

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
activation of protein kinase activity
protein autophosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium ion binding
protein binding
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0