This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01196366_m1
See other HELQ GE Assays ›
Gene Symbol:
Helq
Gene Name:
helicase, POLQ-like
Gene Aliases:
AU021725, D430018E21Rik, Hel308
Chromosome Location:
Chr.5: 100762144 - 100798663 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01196366_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Helq
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081107.1 NP_001074576.1 8 - 9 1818 144
GenBank mRNA AF436846.1 - 6 - 7 800 144
AK052427.1 - 8 - 9 1778 144
BC082601.1 - 8 - 9 1782 30662103 144
BC109169.2 - 8 - 9 1686 144
BC109170.2 - 8 - 9 1691 40048258 144

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA metabolic process
DNA duplex unwinding
nucleotide binding
nucleic acid binding
helicase activity
ATP binding
hydrolase activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0