This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01195890_m1
See other SLC1A1 GE Assays ›
Gene Symbol:
Slc1a1
Gene Name:
solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1
Gene Aliases:
D130048G10Rik, EAAC1, EAAC2, EAAT3, MEAAC1
Chromosome Location:
Chr.19: 28835166 - 28913960 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01195890_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009199.2 NP_033225.1 1 - 2 165 90
GenBank mRNA AK045632.1 - 1 - 2 197 90
BC064797.1 - 1 - 2 187 30029501 90
BC065099.1 - 1 - 2 189 30364069 90
D43797.1 - 1 - 2 171 90
U73521.1 - 1 - 2 165 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000426.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc1a1

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
dicarboxylic acid transport
L-glutamate transport
sodium ion transmembrane transport
cysteine transport
protein homooligomerization
L-glutamate import
D-aspartate import
L-glutamate transmembrane transport
cysteine transmembrane transport
L-glutamate transmembrane transporter activity
protein binding
symporter activity
glutamate:sodium symporter activity
glutamate binding
sodium:dicarboxylate symporter activity
cysteine transmembrane transporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900003945.mm.1 NM_009199.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003945.mm.1 NM_009199.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900003945.mm.1 TC1900000426.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900003943.mm.1 NM_009199.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900003943.mm.1 NM_009199.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900003943.mm.1 TC1900000426.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 338995 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0