This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194802_m1
See other A1CF GE Assays ›
Gene Symbol:
A1cf
Gene Name:
APOBEC1 complementation factor
Gene Aliases:
1810073H04Rik, ACF64, ACF65, ASP, Acf
Chromosome Location:
Chr.19: 31868755 - 31952365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194802_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A1cf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081074.1 NP_001074543.1 1 - 2 106 126
GenBank mRNA AY566862.1 - 1 - 2 103 126
AY566863.1 - 1 - 2 103 126
AY566864.1 - 1 - 2 103 126

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY566862.1, AY566863.1, AY566864.1, NM_001081074.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
mRNA localization resulting in posttranscriptional regulation of gene expression
cytidine to uridine editing
mRNA modification
protein stabilization
nucleotide binding
nucleic acid binding
RNA binding
single-stranded RNA binding
mRNA binding
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004221.mm.1 NM_001081074.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004221.mm.1 NM_001081074.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004221.mm.1 TC1900000461.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004219.mm.1 NM_001081074.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004219.mm.1 NM_001081074.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004219.mm.1 TC1900000461.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004220.mm.1 NM_001081074.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004220.mm.1 NM_001081074.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004220.mm.1 TC1900000461.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 544961 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header