This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01193905_m1
See other ST3GAL3 GE Assays ›
Gene Symbol:
St3gal3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
ST3GalIII, ST3N, Siat3, Siat6
Chromosome Location:
Chr.4: 117932152 - 118134946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01193905_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: St3gal3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161774.2 NP_001155246.1 9 - 10 985 64
NM_001285520.1 NP_001272449.1 9 - 10 1315 64
NM_001285521.1 NP_001272450.1 8 - 9 937 64
NM_009176.5 NP_033202.3 9 - 10 1329 64
GenBank mRNA AK005053.1 - 9 - 10 1274 64
AK013016.1 - 8 - 9 876 64
AK136442.1 - 9 - 10 1266 64
BC006710.1 - 9 - 10 953 3584323 64
X84234.1 - 9 - 10 808 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003383.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene St3gal3

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
sialyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400027270.mm.1 NM_001161774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400027270.mm.1 NM_001285520.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027270.mm.1 NM_001285521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027270.mm.1 NM_009176.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400027270.mm.1 TC0400003383.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400027271.mm.1 NM_001161774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400027271.mm.1 NM_001285520.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027271.mm.1 NM_001285521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027271.mm.1 NM_009176.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400027271.mm.1 TC0400003383.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0