This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01193902_m1
See other ST3GAL3 GE Assays ›
Gene Symbol:
St3gal3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
ST3GalIII, ST3N, Siat3, Siat6
Chromosome Location:
Chr.4: 117932152 - 118134946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01193902_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: St3gal3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161774.2 NP_001155246.1 2 - 3 355 66
NM_001285520.1 NP_001272449.1 2 - 3 685 66
NM_009176.5 NP_033202.3 2 - 3 699 66
GenBank mRNA AK005053.1 - 2 - 3 644 66
AK136442.1 - 2 - 3 636 66
BC006710.1 - 2 - 3 323 3584323 66
BQ951077.1 - 2 - 3 655 66
CX240214.1 - 2 - 3 315 66
X84234.1 - 2 - 3 178 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003383.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene St3gal3

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
sialyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400027284.mm.1 NM_001161774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400027284.mm.1 NM_001285520.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027284.mm.1 NM_001285521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027284.mm.1 NM_009176.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400027284.mm.1 TC0400003383.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400027280.mm.1 NM_001161774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400027280.mm.1 NM_001285520.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027280.mm.1 NM_009176.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400027280.mm.1 TC0400003383.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 826245 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0