Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700003707.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003707.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003722.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003722.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003722.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003726.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003726.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003726.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003727.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003727.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003727.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003732.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003732.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003732.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003733.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003733.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003733.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003724.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003724.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003724.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003734.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003734.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003734.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003728.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003728.mm.1 |
NM_007434.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003728.mm.1 |
TC0700000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003708.mm.1 |
NM_001110208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003708.mm.1 |
TC0700000465.mm.1 |
MTA TC |