This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191904_m1
See other SEC63 GE Assays ›
Gene Symbol:
Sec63
Gene Name:
SEC63-like (S. cerevisiae)
Gene Aliases:
5730478J10Rik, AI649014, AW319215
Chromosome Location:
Chr.10: 42761496 - 42832514 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191904_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec63
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153055.3 NP_694695.3 5 - 6 782 117
GenBank mRNA AK036629.1 - 5 - 6 477 117
AK049737.1 - 5 - 6 473 117
AY024346.1 - 5 - 6 579 117
AY238936.1 - 5 - 6 552 117
BC019366.1 - 5 - 6 546 5066908 117
BC031846.1 - 5 - 6 546 5066822 117
BC059816.1 - 5 - 6 781 117
BC080810.1 - 5 - 6 663 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000386.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec63

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

liver development
SRP-dependent cotranslational protein targeting to membrane
posttranslational protein targeting to membrane
nitrogen compound metabolic process
transport
multicellular organism aging
protein transport
posttranslational protein targeting to membrane, translocation
renal system development
protein transporter activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000002638.mm.1 NM_153055.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002638.mm.1 NM_153055.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000002638.mm.1 TC1000000386.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0