This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191474_g1
See other POLM GE Assays ›
Gene Symbol:
Polm
Gene Name:
polymerase (DNA directed), mu
Gene Aliases:
AI462166, B230309I03Rik, Tdt-N
Chromosome Location:
Chr.11: 5827860 - 5838341 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191474_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017401.2 NP_059097.2 2 - 3 642 133
GenBank mRNA AF176098.1 - 2 - 3 415 133
AJ251804.2 - 2 - 3 386 133
AK137747.1 - 1 - 2 1593 134
BC056213.1 - 2 - 3 434 30024844 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002154.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polm

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
cellular response to DNA damage stimulus
somatic hypermutation of immunoglobulin genes
B cell differentiation
DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
DNA polymerase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100020924.mm.1 NM_017401.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020924.mm.1 NM_017401.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100020924.mm.1 TC1100002154.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100020925.mm.1 NM_017401.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020925.mm.1 NM_017401.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100020925.mm.1 TC1100002154.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0