This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01190305_m1
See other HNRNPF GE Assays ›
Gene Symbol:
Hnrnpf
Gene Name:
heterogeneous nuclear ribonucleoprotein F
Gene Aliases:
4833420I20Rik, AA407306, Hnrpf
Chromosome Location:
Chr.6: 117900340 - 117925622 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01190305_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hnrnpf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166429.1 NP_001159901.1 1 - 2 81 71
GenBank mRNA CK389060.1 - 1 - 2 72 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001166429.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
regulation of RNA splicing
nucleotide binding
nucleic acid binding
RNA binding
single-stranded RNA binding
transcription factor binding
TBP-class protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010492.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010491.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010494.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010482.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010485.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010487.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010493.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010474.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010474.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010474.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010474.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010474.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010489.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010481.mm.1 TC0600001322.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166427.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166428.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166428.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166429.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166429.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_001166432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_133834.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 NM_133834.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010480.mm.1 TC0600001322.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0