This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188857_m1
See other C1RL GE Assays ›
Gene Symbol:
C1rl
Gene Name:
complement component 1, r subcomponent-like
Gene Aliases:
C1r-LP, C1rl1, C1rlp
Chromosome Location:
Chr.6: 124493113 - 124510643 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188857_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C1rl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181344.5 NP_851989.3 5 - 6 714 97
GenBank mRNA AF456428.1 - 4 - 5 598 97
AK038659.1 - 5 - 6 680 97
AK133495.1 - 5 - 6 693 97
AK134207.1 - 5 - 6 715 97
AK162497.1 - 5 - 6 669 97
BC070472.1 - 5 - 6 677 30627947 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene C1rl

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

immune system process
proteolysis
complement activation, classical pathway
innate immune response
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098226 AK162497.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098226 AK133495.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098226 NM_181344.5 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211227 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552962 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474410 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175051 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956916 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432791 AK031713.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432791 AK043666.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344131 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37488 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302623 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158996 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122481 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155830 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106737 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137186 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098226 AK162497.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098226 AK133495.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098226 NM_181344.5 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251367 AK162497.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251367 BC070472.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251367 AK133495.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251367 AK134207.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251367 NM_181344.5 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815621 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815603 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815613 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815594 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815595 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815601 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815599 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815597 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815619 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815584 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815602 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815605 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815622 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815586 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815620 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815607 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815617 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815587 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815624 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815591 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815583 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815581 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815615 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815610 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815588 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815616 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815590 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01437435 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01437438 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106737 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055754 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137186 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01437433 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151872 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055772 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055683 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055729 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055631 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055764 - -
Whole Mouse Genome G4122A A_51_P251367 AK162497.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P251367 BC070472.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P251367 AK133495.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P251367 AK134207.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P251367 NM_181344.5 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104036 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110813 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06055764 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137186 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106737 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151872 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06055683 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06055729 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01437433 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05644225 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05609779 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05609780 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05644224 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05644222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815962 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815565 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815667 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815580 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815627 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05754478 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05121832 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05628890 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815654 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815607 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815619 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815661 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05628882 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815580 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815590 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815632 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815644 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815640 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815656 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05754475 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815637 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815617 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05263382 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05210855 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05126720 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815588 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815601 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05289284 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815627 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815630 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815634 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815603 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05644222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815636 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815592 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815626 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05173830 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815652 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815620 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815655 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05552079 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05628885 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05628889 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05628884 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815628 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815663 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05543073 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815605 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815672 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815633 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05754470 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815650 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05742776 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05322468 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815639 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815676 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05039852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05798443 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05742779 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815641 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05802731 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815586 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05121837 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815610 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24815657 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05658556 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10541671 - -
Affymetrix Mouse Gene 1.0 ST Array 10541676 - -
Affymetrix Mouse Gene 1.0 ST Array 10541674 - -
Affymetrix Mouse Gene 1.0 ST Array 10541672 - -
Affymetrix Mouse Gene 1.0 ST Array 10541673 - -
Affymetrix Mouse Gene 1.0 ST Array 10541677 - -
Affymetrix Mouse Gene 1.0 ST Array 10541675 - -
Affymetrix Mouse Gene 1.1 ST Array 10541670 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 438215 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1690142 - -
Illumina mouse-6 v1.0 expression beadchip 1690142 - -
Illumina mouse-6 v1.0 expression beadchip 6650092 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006650092 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001690142 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255190 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2728796 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2728796 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1255190 - -
mouse-6 v1.1 (Illumina) scl0232371.6_16-S - -
mouse-6 v1.1 (Illumina) scl29523.5.1_4-S - -
Illumina Sentrix mouse-6.v1 bead array 1690142 - -
Illumina Sentrix mouse-6.v1 bead array 6650092 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl29523.5.1_4-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0232371.6_16-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690142 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650092 - -
MouseRef-8 v2 Expression BeadChip ILMN_2728796 - -
MouseRef-8 v2 Expression BeadChip ILMN_1255190 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2728796 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255190 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0