This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188637_m1
See other CNTN4 GE Assays ›
Gene Symbol:
Cntn4
Gene Name:
contactin 4
Gene Aliases:
9630050B05, Axcam, BIG-2
Chromosome Location:
Chr.6: 105677651 - 106700141 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188637_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cntn4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109749.1 NP_001103219.1 18 - 19 2443 74
NM_001109751.1 NP_001103221.1 17 - 18 2531 74
NM_173004.3 NP_766592.2 18 - 19 2443 74
GenBank mRNA AK036262.1 - 18 - 19 2413 74
AK044694.1 - 8 - 9 1190 74
AK173340.1 - 11 - 12 1562 74
BC046975.1 - 15 - 16 2065 74
BC115766.1 - 18 - 19 2333 40060288 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cntn4

Gene Ontology Categories:

Function(s)

cell adhesion
nervous system development
neuron projection development
negative regulation of neuron differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header