This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188519_m1
See other AGO4 GE Assays ›
Gene Symbol:
Ago4
Gene Name:
argonaute RISC catalytic subunit 4
Gene Aliases:
5730550L01Rik, AI481660, Eif2c4
Chromosome Location:
Chr.4: 126489487 - 126533588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188519_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ago4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153177.3 NP_694817.2 11 - 12 1631 79
GenBank mRNA AB081474.1 - 11 - 12 1376 79
AK030018.1 - 11 - 12 1647 79
BC096023.1 - 11 - 12 1413 30607807 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003545.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ago4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA catabolic process
regulation of translation
RNA secondary structure unwinding
gene silencing by RNA
pre-miRNA processing
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
miRNA loading onto RISC involved in gene silencing by miRNA
nucleic acid binding
RNA binding
double-stranded RNA binding
single-stranded RNA binding
miRNA binding
pre-miRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400028878.mm.1 NM_153177.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028878.mm.1 NM_153177.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400028878.mm.1 TC0400003545.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0