This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188420_m1
See other NHSL1 GE Assays ›
Gene Symbol:
Nhsl1
Gene Name:
NHS-like 1
Gene Aliases:
5730409E15Rik, A630035H13Rik, A730096F01, BC013565, D10Bwg0940e, mKIAA1357
Chromosome Location:
Chr.10: 18318558 - 18533892 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188420_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nhsl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163592.1 NP_001157064.1 5 - 6 787 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000131.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000000909.mm.1 NM_001163592.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000909.mm.1 NM_001163592.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000909.mm.1 NM_173390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000909.mm.1 NM_173390.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000000909.mm.1 TC1000000131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000000910.mm.1 NM_001163592.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000910.mm.1 NM_001163592.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000910.mm.1 NM_173390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000910.mm.1 NM_173390.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000000910.mm.1 TC1000000131.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0