This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188213_m1
See other S100PBP GE Assays ›
Gene Symbol:
S100pbp
Gene Name:
S100P binding protein
Gene Aliases:
4930429A08Rik, AI851343, S100pbpr
Chromosome Location:
Chr.4: 129148002 - 129189742 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188213_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100pbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029036.2 NP_083312.1 5 - 6 1190 130
GenBank mRNA AK015229.1 - 5 - 6 1190 130
AK030636.1 - 7 - 8 884 130
AK043160.1 - 3 - 4 194 130
AK046756.1 - 5 - 6 2236 130
AK146310.1 - 5 - 6 1253 130
BC038329.1 - 7 - 8 840 1548746 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003586.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100pbp

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400029194.mm.1 NM_029036.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029194.mm.1 NM_029036.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400029194.mm.1 TC0400003586.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header