Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029221.mm.1 |
TC0400003586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029223.mm.1 |
TC0400003586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029213.mm.1 |
NM_029036.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029213.mm.1 |
NM_029036.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029213.mm.1 |
TC0400003586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029211.mm.1 |
NM_029036.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029211.mm.1 |
NM_029036.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029211.mm.1 |
TC0400003586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029226.mm.1 |
TC0400003586.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029212.mm.1 |
NM_029036.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029212.mm.1 |
NM_029036.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029212.mm.1 |
TC0400003586.mm.1 |
MTA TC |