This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187626_m1
See other SEC14L3 GE Assays ›
Gene Symbol:
Sec14l3
Gene Name:
SEC14-like 3 (S. cerevisiae)
Gene Aliases:
1110069O07Rik
Chromosome Location:
Chr.11: 4064853 - 4078990 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187626_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec14l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029937.2 NP_001025108.1 7 - 8 680 88
GenBank mRNA BC138982.1 - 7 - 8 680 8862004 88
BC138985.1 - 7 - 8 680 8862007 88
BC145485.1 - 7 - 8 723 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000033.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec14l3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100000411.mm.1 NM_001029937.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000411.mm.1 NM_001029937.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000411.mm.1 TC1100000033.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0